Assay Design



Gene Transcripts

Gene Symbol
CLMP
Entrez Gene ID
516565
Gene Name
CXADR like membrane protein
Gene Aliases
ASAM
Chromosome Location
Chr.15: 33689071 - 33794265 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098043.1 NP_001091512.1 5 - 6 987 85
GenBank mRNA BC140573.1 - 5 - 6 987 85

View Full Product Details