Assay Design



Gene Transcripts

Gene Symbol
GUCD1
Entrez Gene ID
517135
Gene Name
guanylyl cyclase domain containing 1
Gene Aliases
C17H22orf13, C22orf13
Chromosome Location
Chr.17: 71503785 - 71515636 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098044.1 NP_001091513.1 4 - 5 491 124
GenBank mRNA BC140595.1 - 4 - 5 491 124

View Full Product Details