Assay Design



Gene Transcripts

Gene Symbol
MPZL3
Entrez Gene ID
521837
Gene Name
myelin protein zero like 3
Gene Aliases
-
Chromosome Location
Chr.15: 28913762 - 28941377 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098051.1 NP_001091520.1 3 - 4 489 56
GenBank mRNA BC140504.1 - 3 - 4 489 56

View Full Product Details