Assay Design



Gene Transcripts

Gene Symbol
TAP1
Entrez Gene ID
524959
Gene Name
transporter 1, ATP binding cassette subfamily B member
Gene Aliases
-
Chromosome Location
Chr.23: 7177596 - 7186556 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098058.1 NP_001091527.1 2 - 3 713 70
GenBank mRNA BC149569.1 - 2 - 3 1014 70

View Full Product Details