Assay Design



Gene Transcripts

Gene Symbol
PITHD1
Entrez Gene ID
506494
Gene Name
PITH domain containing 1
Gene Aliases
C2H1orf128
Chromosome Location
Chr.2: 129122972 - 129130667 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098890.2 NP_001092360.1 4 - 5 474 118
GenBank mRNA BC142400.1 - 4 - 5 476 118

View Full Product Details