Assay Design



Gene Transcripts

Gene Symbol
COPRS
Entrez Gene ID
506999
Gene Name
coordinator of PRMT5 and differentiation stimulator
Gene Aliases
C19H17orf79, COPR5
Chromosome Location
Chr.19: 18255752 - 18260611 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098894.2 NP_001092364.1 2 - 3 213 60
GenBank mRNA BC142058.1 - 2 - 3 214 60

View Full Product Details