Assay Design



Gene Transcripts

Gene Symbol
HPCAL1
Entrez Gene ID
513870
Gene Name
hippocalcin like 1
Gene Aliases
-
Chromosome Location
Chr.11: 87213318 - 87330638 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098964.2 NP_001092434.1 2 - 3 379 85
GenBank mRNA BC142529.1 - 2 - 3 380 85

View Full Product Details