Assay Design



Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
514047
Gene Name
chymotrypsin C
Gene Aliases
bPTLP
Chromosome Location
Chr.16: 52045641 - 52052536 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098965.1 NP_001092435.1 5 - 6 521 67
GenBank mRNA BC134798.1 - 5 - 6 522 67
BC142035.1 - 5 - 6 521 67
BC151506.1 - 5 - 6 522 67

View Full Product Details