Assay Design



Gene Transcripts

Gene Symbol
TEKT3
Entrez Gene ID
522915
Gene Name
tektin 3
Gene Aliases
OMC45
Chromosome Location
Chr.19: 32816854 - 32849899 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099019.2 NP_001092489.1 6 - 7 1165 103
GenBank mRNA BC146148.1 - 6 - 7 1166 103

View Full Product Details