Assay Design



Gene Transcripts

Gene Symbol
CLIC3
Entrez Gene ID
505436
Gene Name
chloride intracellular channel 3
Gene Aliases
-
Chromosome Location
Chr.11: 106039204 - 106043647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100301.2 NP_001093771.1 5 - 6 571 98
GenBank mRNA BC147880.1 - 5 - 6 572 98

View Full Product Details