Assay Design



Gene Transcripts

Gene Symbol
LMOD3
Entrez Gene ID
509978
Gene Name
leiomodin 3
Gene Aliases
-
Chromosome Location
Chr.22: 32417250 - 32434415 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100332.1 NP_001093802.1 3 - 4 1833 121
GenBank mRNA BC149219.1 - 3 - 4 1833 121

View Full Product Details