Assay Design



Gene Transcripts

Gene Symbol
METTL21C
Entrez Gene ID
519447
Gene Name
methyltransferase like 21C
Gene Aliases
C12H13orf39
Chromosome Location
Chr.12: 78920520 - 78931943 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100383.1 NP_001093853.1 3 - 4 698 71
GenBank mRNA BC149196.1 - 3 - 4 698 71

View Full Product Details