Assay Design



Gene Transcripts

Gene Symbol
PF4
Entrez Gene ID
507790
Gene Name
platelet factor 4
Gene Aliases
-
Chromosome Location
Chr.6: 88924248 - 88925219 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101062.1 NP_001094532.1 2 - 3 339 104
GenBank mRNA BC149422.1 - 2 - 3 339 104

View Full Product Details