Assay Design



Gene Transcripts

Gene Symbol
TSPYL4
Entrez Gene ID
508104
Gene Name
TSPY like 4
Gene Aliases
-
Chromosome Location
Chr.9: 34447805 - 34451911 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101064.2 NP_001094534.1 1 - 1 3555 139
GenBank mRNA BC151251.1 - 1 - 1 3556 139

View Full Product Details