Assay Design



Gene Transcripts

Gene Symbol
TSPAN4
Entrez Gene ID
505288
Gene Name
tetraspanin 4
Gene Aliases
-
Chromosome Location
Chr.29: 50514095 - 50530507 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101852.1 NP_001095322.1 4 - 5 750 83
GenBank mRNA BC150101.1 - 4 - 5 750 83

View Full Product Details