Assay Design



Gene Transcripts

Gene Symbol
OLFM1
Entrez Gene ID
507002
Gene Name
olfactomedin 1
Gene Aliases
-
Chromosome Location
Chr.11: 105560462 - 105581643 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101879.2 NP_001095349.1 2 - 3 411 113
GenBank mRNA BC149341.1 - 2 - 3 412 113

View Full Product Details