Assay Design



Gene Transcripts

Gene Symbol
TRAPPC13
Entrez Gene ID
511108
Gene Name
trafficking protein particle complex 13
Gene Aliases
C20H5orf44
Chromosome Location
Chr.20: 13902562 - 13938722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101940.1 NP_001095410.1 2 - 3 374 130
GenBank mRNA BC151376.1 - 2 - 3 374 130

View Full Product Details