Assay Design



Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
513753
Gene Name
endothelin 3
Gene Aliases
-
Chromosome Location
Chr.13: 57061094 - 57088993 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101979.1 NP_001095449.1 1 - 2 242 65
GenBank mRNA BC148051.1 - 1 - 2 242 65

View Full Product Details