Assay Design



Gene Transcripts

Gene Symbol
NAB1
Entrez Gene ID
516781
Gene Name
NGFI-A binding protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 5634336 - 5683750 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102039.1 NP_001095509.1 4 - 5 1099 97
GenBank mRNA BC151355.1 - 4 - 5 1099 97

View Full Product Details