Assay Design



Gene Transcripts

Gene Symbol
RUBCNL
Entrez Gene ID
517231
Gene Name
rubicon like autophagy enhancer
Gene Aliases
C12H13orf18, KIAA0226L
Chromosome Location
Chr.12: 16369971 - 16410652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102041.2 NP_001095511.1 9 - 10 1479 68
GenBank mRNA BC151662.1 - 9 - 10 1480 68

View Full Product Details