Assay Design



Gene Transcripts

Gene Symbol
CLHC1
Entrez Gene ID
517621
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
C11H2orf63
Chromosome Location
Chr.11: 37935177 - 37970181 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102042.2 NP_001095512.2 1 - 2 43 87
GenBank mRNA BC148948.1 - 1 - 2 43 87

View Full Product Details