Assay Design



Gene Transcripts

Gene Symbol
CLEC12A
Entrez Gene ID
508740
Gene Name
C-type lectin domain family 12 member A
Gene Aliases
-
Chromosome Location
Chr.5: 99947392 - 99963231 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105345.2 NP_001098815.1 3 - 4 480 145
GenBank mRNA BC151519.1 - 3 - 4 481 145

View Full Product Details