Assay Design



Gene Transcripts

Gene Symbol
DTX3
Entrez Gene ID
524779
Gene Name
deltex E3 ubiquitin ligase 3
Gene Aliases
-
Chromosome Location
Chr.5: 55880126 - 55885446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105393.1 NP_001098863.1 1 - 2 60 85
GenBank mRNA BC153272.1 - 1 - 2 60 85

View Full Product Details