Assay Design



Gene Transcripts

Gene Symbol
RSPO1
Entrez Gene ID
511675
Gene Name
R-spondin 1
Gene Aliases
-
Chromosome Location
Chr.3: 108267191 - 108288925 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105621.1 NP_001099091.1 4 - 5 988 62
GenBank mRNA BC149052.1 - 4 - 5 988 62
BT030732.1 - 2 - 3 313 62

View Full Product Details