Assay Design



Gene Transcripts

Gene Symbol
CYP2C19
Entrez Gene ID
510380
Gene Name
cytochrome P450, family 2, subfamily C, polypeptide 19
Gene Aliases
-
Chromosome Location
Chr.26: 16473502 - 16520973 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109792.2 NP_001103262.1 5 - 6 838 106
GenBank mRNA AY265992.1 - 1 - 2 66 106
BC153845.1 - 5 - 6 840 106

View Full Product Details