Assay Design



Gene Transcripts

Gene Symbol
AQP11
Entrez Gene ID
510038
Gene Name
aquaporin 11
Gene Aliases
-
Chromosome Location
Chr.29: 18378215 - 18393790 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110069.1 NP_001103539.1 2 - 3 774 140
GenBank mRNA BC120161.1 - 2 - 3 774 140

View Full Product Details