Assay Design



Gene Transcripts

Gene Symbol
CTSW
Entrez Gene ID
510967
Gene Name
cathepsin W
Gene Aliases
-
Chromosome Location
Chr.29: 43995837 - 43999310 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110070.1 NP_001103540.1 2 - 3 174 71
GenBank mRNA BC113312.1 - 2 - 3 174 71

View Full Product Details