Assay Design



Gene Transcripts

Gene Symbol
PTS
Entrez Gene ID
506860
Gene Name
6-pyruvoyltetrahydropterin synthase
Gene Aliases
-
Chromosome Location
Chr.15: 22587044 - 22624477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113537.1 NP_001107009.1 1 - 2 127 93
GenBank mRNA CB455463.1 - 1 - 2 132 93

View Full Product Details