Assay Design



Gene Transcripts

Gene Symbol
KANSL2
Entrez Gene ID
540194
Gene Name
KAT8 regulatory NSL complex subunit 2
Gene Aliases
C12orf41, C5H12orf41, C5HANSL2
Chromosome Location
Chr.5: 31099668 - 31117925 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024571.2 NP_001019742.2 4 - 5 589 76
GenBank mRNA BC111221.1 - 4 - 5 589 76
BT021786.1 - 3 - 4 528 76

View Full Product Details