Assay Design



Gene Transcripts

Gene Symbol
KANSL2
Entrez Gene ID
540194
Gene Name
KAT8 regulatory NSL complex subunit 2
Gene Aliases
C12orf41, C5H12orf41, C5HANSL2
Chromosome Location
Chr.5: 31099668 - 31117925 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024571.2 NP_001019742.2 6 - 7 916 59
GenBank mRNA BC111221.1 - 6 - 7 916 59
BT021786.1 - 5 - 6 855 59

View Full Product Details