Assay Design



Gene Transcripts

Gene Symbol
VPS35L
Entrez Gene ID
534058
Gene Name
VPS35 endosomal protein sorting factor like
Gene Aliases
C16orf62, C25H16orf62
Chromosome Location
Chr.25: 17159011 - 17297111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046368.2 NP_001039833.2 12 - 13 1041 114
GenBank mRNA DV883546.1 - 9 - 10 723 114

View Full Product Details