Assay Design



Gene Transcripts

Gene Symbol
LDHC
Entrez Gene ID
537256
Gene Name
lactate dehydrogenase C
Gene Aliases
-
Chromosome Location
Chr.29: 26253200 - 26286422 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046422.2 NP_001039887.2 3 - 4 527 109
NM_001113249.1 NP_001106720.1 3 - 4 527 109
GenBank mRNA BC105361.1 - 3 - 4 510 109
DT830273.1 - 3 - 4 313 109
DT847039.1 - 3 - 4 531 109

View Full Product Details