Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076098.2 | NP_001069566.2 | 2 - 3 | 348 | 61 |
GenBank mRNA | BC120190.1 | - | 2 - 3 | 348 | 61 |
DT830787.1 | - | 2 - 3 | 322 | 61 | |
EE246344.1 | - | 2 - 3 | 348 | 61 |