Assay Design



Gene Transcripts

Gene Symbol
TFB1M
Entrez Gene ID
533438
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.9: 91865417 - 91932613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076896.2 NP_001070364.1 1 - 2 188 75
NM_001114610.1 NP_001108082.1 1 - 2 188 75
GenBank mRNA BC109841.1 - 1 - 2 176 75
DT811342.1 - 1 - 2 181 75
DY454254.1 - 1 - 2 192 75

View Full Product Details