Assay Design



Gene Transcripts

Gene Symbol
LRRC17
Entrez Gene ID
777690
Gene Name
leucine rich repeat containing 17
Gene Aliases
-
Chromosome Location
Chr.4: 44244168 - 44279713 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078150.2 NP_001071618.1 2 - 3 1103 115
GenBank mRNA BC113338.1 - 2 - 3 949 115
BT030519.1 - 2 - 3 1103 115

View Full Product Details