Assay Design



Gene Transcripts

Gene Symbol
USP12
Entrez Gene ID
525655
Gene Name
ubiquitin specific peptidase 12
Gene Aliases
USP12P1
Chromosome Location
Chr.12: 32655257 - 32715132 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098059.1 NP_001091528.1 6 - 7 799 133
GenBank mRNA BT030596.2 - 6 - 7 797 133

View Full Product Details