Assay Design



Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
525916
Gene Name
BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.21: 24995268 - 25047183 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098060.2 NP_001091529.1 4 - 5 955 71
GenBank mRNA BC140696.1 - 4 - 5 956 71

View Full Product Details