Assay Design



Gene Transcripts

Gene Symbol
YARS2
Entrez Gene ID
536956
Gene Name
tyrosyl-tRNA synthetase 2
Gene Aliases
-
Chromosome Location
Chr.5: 77131736 - 77158405 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098088.2 NP_001091557.1 4 - 5 1365 98
GenBank mRNA BC140541.1 - 4 - 5 1366 98

View Full Product Details