Assay Design



Gene Transcripts

Gene Symbol
ART1
Entrez Gene ID
539042
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.15: 51351999 - 51388933 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098098.1 NP_001091567.1 1 - 2 39 66
GenBank mRNA BC140591.1 - 1 - 2 39 66

View Full Product Details