Assay Design



Gene Transcripts

Gene Symbol
NCK2
Entrez Gene ID
526430
Gene Name
NCK adaptor protein 2
Gene Aliases
-
Chromosome Location
Chr.11: 45918920 - 46034068 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099036.1 NP_001092506.1 1 - 2 77 66
GenBank mRNA BC146083.1 - 1 - 2 77 66

View Full Product Details