Assay Design



Gene Transcripts

Gene Symbol
TNFRSF4
Entrez Gene ID
528782
Gene Name
TNF receptor superfamily member 4
Gene Aliases
-
Chromosome Location
Chr.16: 51285833 - 51288677 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099043.2 NP_001092513.1 3 - 4 405 134
GenBank mRNA BC142115.1 - 3 - 4 406 134

View Full Product Details