Assay Design



Gene Transcripts

Gene Symbol
MAPK7
Entrez Gene ID
537703
Gene Name
mitogen-activated protein kinase 7
Gene Aliases
ERK-5
Chromosome Location
Chr.19: 34085200 - 34091163 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099080.2 NP_001092550.1 1 - 2 610 114
GenBank mRNA BC142510.1 - 1 - 2 611 114

View Full Product Details