Assay Design



Gene Transcripts

Gene Symbol
RCE1
Entrez Gene ID
539539
Gene Name
Ras converting CAAX endopeptidase 1
Gene Aliases
FACE-2
Chromosome Location
Chr.29: 44857553 - 44860678 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099097.1 NP_001092567.1 2 - 3 303 58
GenBank mRNA BC146167.1 - 2 - 3 303 58

View Full Product Details