Assay Design



Gene Transcripts

Gene Symbol
IGFLR1
Entrez Gene ID
617594
Gene Name
IGF like family receptor 1
Gene Aliases
TMEM149
Chromosome Location
Chr.18: 46454241 - 46456880 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099167.2 NP_001092637.1 2 - 3 412 98
GenBank mRNA BC142051.1 - 2 - 3 413 98

View Full Product Details