Assay Design



Gene Transcripts

Gene Symbol
FKRP
Entrez Gene ID
539701
Gene Name
fukutin related protein
Gene Aliases
-
Chromosome Location
Chr.18: 53849380 - 53859525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099714.1 NP_001093184.1 2 - 3 89 81
GenBank mRNA BC142286.1 - 2 - 3 89 81

View Full Product Details