Assay Design



Gene Transcripts

Gene Symbol
LSMEM1
Entrez Gene ID
613798
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C4H7orf53, C7orf53
Chromosome Location
Chr.4: 55849062 - 55863182 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099718.2 NP_001093188.1 1 - 2 367 60
GenBank mRNA BC142316.1 - 1 - 2 368 60

View Full Product Details