Assay Design



Gene Transcripts

Gene Symbol
MAT2A
Entrez Gene ID
527403
Gene Name
methionine adenosyltransferase 2A
Gene Aliases
-
Chromosome Location
Chr.11: 49465580 - 49472218 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101131.1 NP_001094601.1 6 - 7 870 62
GenBank mRNA BT030708.1 - 6 - 7 870 62

View Full Product Details