Assay Design



Gene Transcripts

Gene Symbol
SPACA9
Entrez Gene ID
539447
Gene Name
sperm acrosome associated 9
Gene Aliases
C11H9orf9
Chromosome Location
Chr.11: 102911359 - 102923690 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101193.2 NP_001094663.1 3 - 4 478 54
GenBank mRNA BC146122.1 - 3 - 4 479 54

View Full Product Details