Assay Design



Gene Transcripts

Gene Symbol
AKIRIN1
Entrez Gene ID
615039
Gene Name
akirin 1
Gene Aliases
-
Chromosome Location
Chr.3: 106986683 - 106998848 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101236.1 NP_001094706.1 2 - 3 467 88
GenBank mRNA BC151342.1 - 2 - 3 467 88

View Full Product Details