Assay Design



Gene Transcripts

Gene Symbol
KRTAP4-7
Entrez Gene ID
618486
Gene Name
keratin associated protein 4-7
Gene Aliases
-
Chromosome Location
Chr.19: 41318527 - 41319608 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101273.1 NP_001094743.1 1 - 1 1046 112
GenBank mRNA BC151711.1 - 1 - 1 1046 112

View Full Product Details